1-Brom-2,5-bis(trifluormethyl)benzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Bis(trifluoromethyl)bromobenzene, 2-Bromo-1,4-bis(trifluoromethyl)benzene

M-Clarity™ Qualitätsstufe = MQ100

438782-1G
SIAL438782-1GEA 36.3 EUR
SIAL438782-1G
1-Brom-2,5-bis(trifluormethyl)benzol, Sigma-Aldrich®
1-Brom-2,5-bis(trifluormethyl)benzol
Formel: C₈H₃BrF₆
Molecular Weight: 293,01 g/mol
MDL: MFCD03094336
CAS-Nummer: 7617-93-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR