Yttrium(III)acetylacetonat Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tris(2,4-pentandionato-O,O')yttrium(III) Hydrat, Yttrium(III)-2,4-pentandionat Hydrat, Tri(2,4-pentandionato-O,O')yttrium(III) Hydrat, Yttrium(III)-tris(2,4-pentandionato-O,O') Hydrat

M-Clarity™ Qualitätsstufe = MQ100

438790-5G
SIAL438790-5GEA 135 EUR
SIAL438790-5G
Yttrium(III)acetylacetonat Hydrat, Sigma-Aldrich®
Yttrium(III)acetylacetonat Hydrat
Formel: C₁₅H₂₁O₆Y·H₂O
Molecular Weight: 404,25 g/mol
MDL: MFCD00013509
CAS-Nummer: 207801-29-4
EINECS: 239-607-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR