Hexadecyltrimethylammoniumhydroxid 10% (w/w) in Wasser, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Cetyltrimethylammonium Hydroxide

M-Clarity™ Qualitätsstufe = MQ200

439231-25ML
SIAL439231-25MLEA 136 EUR
SIAL439231-25ML
Hexadecyltrimethylammoniumhydroxid 10% (w/w) in Wasser, Sigma-Aldrich®
Hexadecyltrimethylammonium hydroxide
Formel: C₁₉H₄₃NO
Molecular Weight: 301.56 g/mol
Dichte: 0.988 g/ml (25 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00041921
CAS-Nummer: 505-86-2
UN: 3267
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR