1-Fluor-2,4,6-trimethylpyridiniumtetrafluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Fluoro-2,4,6-trimethylpyridinium Tetrafluoroborate

M-Clarity™ Qualitätsstufe = MQ100

439312-1G
SIAL439312-1GEA 58.5 EUR
SIAL439312-1G
1-Fluor-2,4,6-trimethylpyridiniumtetrafluoroborat, Sigma-Aldrich®
1-Fluoro-2,4,6-trimethylpyridiniumtetrafluoroborate
Formel: C₈H₁₁BF₅N
Molecular Weight: 226,98 g/mol
MDL: MFCD00153182
CAS-Nummer: 109705-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR