4-(Trifluormethyl)phenylboronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(4-(Trifluormethyl)phenyl)metaborsäure, Dihydroxy(4-(trifluormethyl)phenyl)boran, 4-(Trifluormethyl)benzolboronsäure
M-Clarity™ Qualitätsstufe = MQ100
439320-1G
439320-5G
SIAL439320-1GEA
65.7
EUR
SIAL439320-1G
SIAL439320-5G
4-(Trifluormethyl)phenylboronsäure, Sigma-Aldrich®
4-(Trifluormethyl)phenylboronsäure
Formel:
C₇H₆BF₃O₂ Molecular Weight: 189.93 g/mol |
MDL:
MFCD00151855 CAS-Nummer: 128796-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...