4-(Trifluormethyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-(Trifluormethyl)phenyl)metaborsäure, Dihydroxy(4-(trifluormethyl)phenyl)boran, 4-(Trifluormethyl)benzolboronsäure

M-Clarity™ Qualitätsstufe = MQ100

439320-1G 439320-5G
SIAL439320-1GEA 65.7 EUR
SIAL439320-1G SIAL439320-5G
4-(Trifluormethyl)phenylboronsäure, Sigma-Aldrich®
4-(Trifluormethyl)phenylboronsäure
Formel: C₇H₆BF₃O₂
Molecular Weight: 189.93 g/mol
MDL: MFCD00151855
CAS-Nummer: 128796-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR