(4-Hydroxyphenoxy)essigsäure, Sigma-Aldrich®

Lieferant: Merck

439398-5G
SIAL439398-5GEA 49 EUR
SIAL439398-5G
(4-Hydroxyphenoxy)essigsäure, Sigma-Aldrich®
(4-Hydroxyphenoxy)essigsäure
Formel: (C₆H₄(OH))OCH₂COOH
Molecular Weight: 168,15 g/mol
MDL: MFCD00014362
CAS-Nummer: 1878-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR