3-Hydroxy-2,4,6-tribrombenzoesäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,4,6-Tribromo-3-hydroxybenzoic acid
M-Clarity™ Qualitätsstufe = MQ100
439533-5G
SIAL439533-5GEA
45.4
EUR
SIAL439533-5G
3-Hydroxy-2,4,6-tribrombenzoesäure, Sigma-Aldrich®
3-Hydroxy-2,4,6-tribromobenzoic acid
Formel:
C₇H₃Br₃O₃ Molecular Weight: 374,81 g/mol Siedepunkt: 396 °C (1013 hPa) Schmelzpunkt: 146 °C |
MDL:
MFCD00055557 CAS-Nummer: 14348-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...