5-(2-Bromphenyl)-1H-tetrazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
5-(2-Bromophenyl)tetrazole
M-Clarity™ Qualitätsstufe = MQ100
439541-1G
SIAL439541-1GEA
93.2
EUR
SIAL439541-1G
5-(2-Bromphenyl)-1H-tetrazol, Sigma-Aldrich®
5-(2-Bromphenyl)-1H-tetrazol
Formel:
C₇H₅BrN₄ Molecular Weight: 225.05 g/mol Schmelzpunkt: 183…184 °C |
MDL:
MFCD00151799 CAS-Nummer: 73096-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...