Ytterbium(III)chlorid, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

439614-5G
SIAL439614-5GEA 137 EUR
SIAL439614-5G
Ytterbium(III)chlorid, Sigma-Aldrich®
Ytterbium(III)chlorid
Formel: Cl₃Yb
Molecular Weight: 279,4 g/mol
Schmelzpunkt: 860 °C
MDL: MFCD00011288
CAS-Nummer: 10361-91-8
EINECS: 233-800-5
Merck Index: 13,10014

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR