Mangan(II)iodid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Manganous iodide
M-Clarity™ Qualitätsstufe = MQ100
439738-2G
SIAL439738-2GEA
168
EUR
SIAL439738-2G
Mangan(II)iodid, Sigma-Aldrich®
Mangan(II)iodid
Formel:
I₂Mn Molecular Weight: 308,75 g/mol Schmelzpunkt: 638 °C Dichte: 5,04 g/cm³ (20 °C) |
MDL:
MFCD00016225 CAS-Nummer: 7790-33-2 EINECS: 232-201-6 Merck Index: 12,05772 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...