3-(4-Chlorbenzoyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

439924-25G
SIAL439924-25GEA 90.8 EUR
SIAL439924-25G
3-(4-Chlorbenzoyl)propionsäure, Sigma-Aldrich®
3-(4-Chlorbenzoyl)propionsäure
Formel: C₁₀H₉ClO₃
MDL: MFCD00002794
CAS-Nummer: 3984-34-7
EINECS: 223-627-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR