Methyl-2-isocyanatobenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(Methoxycarbonyl)phenyl isocyanate

M-Clarity™ Qualitätsstufe = MQ100

439983-5G
SIAL439983-5GEA 111 EUR
SIAL439983-5G
Methyl-2-isocyanatobenzoat, Sigma-Aldrich®
Methyl-2-isocyanatobenzoat
Formel: C₉H₇NO₃
MDL: MFCD00013848
CAS-Nummer: 1793-07-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR