2-Vinyl-1,3-dioxolan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

440353-5ML
SIAL440353-5MLEA 151 EUR
SIAL440353-5ML
2-Vinyl-1,3-dioxolan, Sigma-Aldrich®
2-Vinyl-1,3-dioxolan
Formel: C₅H₈O₂
CAS-Nummer: 3984-22-3
EINECS: 223-626-8
UN: 2929
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR