N-(2-Hydroxyethyl)-2,2,2-trifluoracetamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2,2-Trifluoro-N-(2-hydroxyethyl)acetamide

440507-25ML
SIAL440507-25MLEA 176 EUR
SIAL440507-25ML
N-(2-Hydroxyethyl)-2,2,2-trifluoracetamid, Sigma-Aldrich®
N-(2-Hydroxyethyl)-2,2,2-trifluoracetamid
Formel: C₄H₆F₃NO₂
Molecular Weight: 157,0912 g/mol
MDL: MFCD00216653
CAS-Nummer: 6974-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR