Ethyltriethoxysilan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Triethoxy(ethyl)silan
M-Clarity™ Qualitätsstufe = MQ100
440574-50ML
SIAL440574-50MLEA
87.4
EUR
SIAL440574-50ML
Ethyltriethoxysilan, Sigma-Aldrich®
Ethyltriethoxysilan
Formel:
C₈H₂₀O₃Si Molecular Weight: 192,33 g/mol Siedepunkt: 158 °C (1013 hPa) Dichte: 0,9207 g/cm³ (0 °C) |
MDL:
MFCD00026754 CAS-Nummer: 78-07-9 EINECS: 201-080-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...