2,3,4,5,6-Pentamethylbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

440620-1G
SIAL440620-1GEA 37.7 EUR
SIAL440620-1G
2,3,4,5,6-Pentamethylbenzylalkohol, Sigma-Aldrich®
2,3,4,5,6-Pentamethylbenzylalkohol
Formel: C₁₂H₁₈O
Molecular Weight: 178,27 g/mol
MDL: MFCD00014421
CAS-Nummer: 484-66-2
EINECS: 207-609-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR