Strontiumisopropoxid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Strontium i-propoxide
M-Clarity™ Qualitätsstufe = MQ100
440698-5G
SIAL440698-5GEA
151
EUR
SIAL440698-5G
Strontiumisopropoxid, Sigma-Aldrich®
Strontiumisopropoxid
Formel:
C₆H₁₄O₂Sr Molecular Weight: 205,79 g/mol |
MDL:
MFCD00050487 CAS-Nummer: 88863-33-6 UN: 3205 ADR: 4.2,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...