4-Thioxo-3,4-dihydro-2(1H)-pyrimidinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxy-4-mercaptopyrimidin

M-Clarity™ Qualitätsstufe = MQ200

440736-1G
SIAL440736-1GEA 121 EUR
SIAL440736-1G
4-Thioxo-3,4-dihydro-2(1H)-pyrimidinon, Sigma-Aldrich®
4-Thiouracil
Formel: C₄H₄N₂OS
Molecular Weight: 128,15 g/mol
MDL: MFCD00090842
CAS-Nummer: 591-28-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR