N-Acetyl-L-leucin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Ac-Leu-OH, Ac-Leu-OH

M-Clarity™ Qualitätsstufe = MQ100

441511-25G
SIAL441511-25GEA 84.4 EUR
SIAL441511-25G
N-Acetyl-L-leucin, Sigma-Aldrich®
N-Acetyl-L-leucin
Formel: C₈H₁₅NO₃
Molecular Weight: 173,2 g/mol
MDL: MFCD00065131
CAS-Nummer: 1188-21-2
EINECS: 214-706-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR