3-Ethoxyphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

441635-1G
SIAL441635-1GEA 28.1 EUR
SIAL441635-1G
3-Ethoxyphenylboronsäure, Sigma-Aldrich®
3-Ethoxyphenylboronsäure
Formel: C₈H₁₁BO₃
Molecular Weight: 165,98 g/mol
MDL: MFCD00274219
CAS-Nummer: 90555-66-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR