3-Formylphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Boronobenzaldehyd, Dihydroxy(3-formylphenyl)boran, (3-Formylphenyl)metaborsäure, 3-Formylbenzolboronsäure

M-Clarity™ Qualitätsstufe = MQ100

441651-5G
SIAL441651-5GEA 33.4 EUR
SIAL441651-5G
3-Formylphenylboronsäure, Sigma-Aldrich®
3-Formylphenylboronsäure
Formel: C₇H₇BO₃
Molecular Weight: 149,94 g/mol
MDL: MFCD00161356
CAS-Nummer: 87199-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR