3-Iodphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

441678-5G
SIAL441678-5GEA 147 EUR
SIAL441678-5G
3-Iodphenylboronsäure, Sigma-Aldrich®
3-Iodphenylboronsäure
Formel: C₆H₆BIO₂
MDL: MFCD01319015
CAS-Nummer: 221037-98-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR