Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] ≥98% (durch TLC), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Tetrakis[methylene-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]methane, Antioxidant 1010, 2,2-Bis(((3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl)oxy)methyl)propane-1,3-diyl bis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate)
M-Clarity™ Qualitätsstufe = MQ200
441783-100G
SIAL441783-100GEA
63.6
EUR
SIAL441783-100G
Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] ≥98% (durch TLC), Sigma-Aldrich®
Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]
Hindered phenolic antioxidant for polymers.
Formel:
C₇₃H₁₀₈O₁₂ Schmelzpunkt: 115…118 °C (Zers., Lit.) |
MDL:
MFCD00059345 CAS-Nummer: 6683-19-8 EINECS: 229-722-6 |
Spezifikation Testergebnisse
Purity (TLC) | ≥98% |
Infrared spectrum | Conforms to structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...