Indium(III)trifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Indium(III) trifluoromethanesulphonate, Indium triflate

M-Clarity™ Qualitätsstufe = MQ100

442151-25G 442151-5G
SIAL442151-25GEA 368 EUR
SIAL442151-25G SIAL442151-5G
Indium(III)trifluormethansulfonat, Sigma-Aldrich®
Indium(III)trifluormethansulfonat
Formel: C₃F₉InO₉S₃
MDL: MFCD00144478
CAS-Nummer: 128008-30-0
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR