Kupfer(II)-ethylendiamin 1.0 M in Wasser, Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

442305-1L
SIAL442305-1LEA 76.1 EUR
SIAL442305-1L
Kupfer(II)-ethylendiamin 1.0 M in Wasser, Sigma-Aldrich®
Kupfer(II)-ethylendiamin

It is used to evaluate cation exchange capacity (CEC) of clays by copper absorption.

Formel: Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
Dichte: 1,086 g/ml (25 °C)
MDL: MFCD00274628
CAS-Nummer: 14552-35-3
UN: 1761
ADR: 8 (6.1),II

Bestellen

Spezifikation Testergebnisse

Colour Dark Blue
Form Liquid
Infrared Spectrum Conforms to Structure
Purity titration by NATH 0,95 - 1,10 M
Concentration 1,0 M in H₂O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR