Lithiumdihydrogenorthophosphat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Lithiumphosphat monobasisch, Lithiumdihydrogenphosphat

442682-500G-A
SIAL442682-500G-AEA 114 EUR
SIAL442682-500G-A
Lithiumdihydrogenorthophosphat, Sigma-Aldrich®
Lithium dihydrogen orthophosphate
Formel: H₂LiO₄P
Molecular Weight: 103,93 g/mol
MDL: MFCD00016188
CAS-Nummer: 13453-80-0
EINECS: 236-633-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR