2-Hydroxy-5-(trifluormethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-(Trifluormethyl)pyridin-2-ol

M-Clarity™ Qualitätsstufe = MQ100

442801-5G-A
SIAL442801-5G-AEA 56.3 EUR
SIAL442801-5G-A
2-Hydroxy-5-(trifluormethyl)pyridin, Sigma-Aldrich®
2-Hydroxy-5-(trifluoromethyl)pyridine
Formel: C₆H₄F₃NO
Molecular Weight: 163,1 g/mol
MDL: MFCD00042315
CAS-Nummer: 33252-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR