AmberChrom™ chloride form 1X8 20-50 mesh, Supelco®

Lieferant: Merck

44324-100G
SIAL44324-100GEA 243 EUR
SIAL44324-100G
AmberChrom™ chloride form 1X8 20-50 mesh, Supelco®
AmberChrom™ chloride form
Formel: (C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x
CAS-Nummer: 69011-19-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR