(S)-3-(Boc-amino)-3-(4-cyanphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-3-((tert-Butoxycarbonyl)amino)-3-(4-cyanophenyl)propanoic acid

44376-500MG-F
SIAL44376-500MG-FEA 350 EUR
SIAL44376-500MG-F
(S)-3-(Boc-amino)-3-(4-cyanphenyl)propionsäure, Sigma-Aldrich®
(S)-3-(Boc-amino)-3-(4-cyanphenyl)propionsäure
Formel: C₁₅H₁₈N₂O₄
Molecular Weight: 290,3144 g/mol
MDL: MFCD03427930
CAS-Nummer: 500770-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR