4-Chlor-2,5-difluorbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

443824-10G
SIAL443824-10GEA 49.2 EUR
SIAL443824-10G
4-Chlor-2,5-difluorbenzoesäure, Sigma-Aldrich®
4-Chloro-2,5-difluorobenzoic acid
Formel: C₇H₃ClF₂O₂
Molecular Weight: 192,55 g/mol
Schmelzpunkt: 153…158 °C
MDL: MFCD00077480
CAS-Nummer: 132794-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR