2,4,6-Tris(trifluormethyl)-1,3,5-triazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4,6-Tris(trifluormethyl)-s-triazin

M-Clarity™ Qualitätsstufe = MQ100

443980-5G
SIAL443980-5GEA 129 EUR
SIAL443980-5G
2,4,6-Tris(trifluormethyl)-1,3,5-triazin, Sigma-Aldrich®
2,4,6-Tris(trifluormethyl)-1,3,5-triazin
Formel: C₆F₉N₃
Molecular Weight: 285,07 g/mol
MDL: MFCD00042436
CAS-Nummer: 368-66-1
EINECS: 206-709-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR