3-Ethyl-3-oxetanmethanol ≥96%, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

444197-10G
SIAL444197-10GEA 74.8 EUR
SIAL444197-10G
3-Ethyl-3-oxetanmethanol ≥96%, Sigma-Aldrich®
3-Ethyl-3-oxetanmethanol
Formel: C₆H₁₂O₂
MDL: MFCD00190143
CAS-Nummer: 3047-32-3
EINECS: 221-254-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR