5-Amino-2,4,6-triiodisophthalsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Amino-2,4,6-triiodoisophthalic Acid

M-Clarity™ Qualitätsstufe = MQ100

444367-50G
SIAL444367-50GEA 135 EUR
SIAL444367-50G
5-Amino-2,4,6-triiodisophthalsäure, Sigma-Aldrich®
5-Amino-2,4,6-triiodisophthalsäure
Formel: C₈H₄I₃NO₄
Molecular Weight: 558.84 g/mol
MDL: MFCD00190167
CAS-Nummer: 35453-19-1
EINECS: 252-575-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR