AmberChrom™ hydrogen form 50W X8 50-100 mesh, stark azide, Supelco®

Lieferant: Merck

44509-100G
SIAL44509-100GEA 269 EUR
SIAL44509-100G
AmberChrom™ hydrogen form 50W X8 50-100 mesh, stark azide, Supelco®
AmberChrom™ hydrogen form
MDL: MFCD00132722
CAS-Nummer: 69011-22-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR