1,1,2,2-Tetramethyldisilan, Sigma-Aldrich®

Lieferant: Merck

445126-1G
SIAL445126-1GEA 227 EUR
SIAL445126-1G
1,1,2,2-Tetramethyldisilan, Sigma-Aldrich®
1,1,2,2-Tetramethyldisilane
Formel: C₄H₁₄Si₂
Molecular Weight: 118,33 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 814-98-2
EINECS: 212-416-1
UN: 1993
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR