2-(Allyloxy)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Salicylaldehyde Allyl Ether, 2-Allyloxybenzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

445509-5ML
SIAL445509-5MLEA 94.1 EUR
SIAL445509-5ML
2-(Allyloxy)benzaldehyd, Sigma-Aldrich®
2-(Allyloxy)benzaldehyd
Formel: C₁₀H₁₀O₂
Molecular Weight: 162,19 g/mol
MDL: MFCD00014130
CAS-Nummer: 28752-82-1
EINECS: 249-198-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR