Hafnium(IV)-tert-butanolat ≥99,99% (Metall-Basis ausgeschlossen Zr) (max. 0,2% Zr), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Hafnium(IV) t-butoxide, Hafnium(IV)-tert-butanolate

M-Clarity™ Qualitätsstufe = MQ100

445541-5G
SIAL445541-5GEA 378 EUR
SIAL445541-5G
Hafnium(IV)-tert-butanolat ≥99,99% (Metall-Basis ausgeschlossen Zr) (max. 0,2% Zr), Sigma-Aldrich®
Hafnium(IV)-tert-butanolat
Formel: C₁₆H₃₆HfO₄
Molecular Weight: 470.94 g/mol
MDL: MFCD00070458
CAS-Nummer: 2172-02-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR