Polyethylenglycoldimethylether, M.W. 2,000, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Poly(ethylene glycol) dimethyl ether Mn 250
M-Clarity™ Qualitätsstufe = MQ200
445908-250G
445908-50G
SIAL445908-250GEA
291
EUR
SIAL445908-250G
SIAL445908-50G
Polyethylenglycoldimethylether, M.W. 2,000, Sigma-Aldrich®
Polyethylenglycoldimethylether
Formel:
H₃C(OCH₂CH₂)nOCH₃ |
MDL:
MFCD00147972 CAS-Nummer: 24991-55-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...