2-(p-Toluolsulfonyl)ethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Tosylethanol

M-Clarity™ Qualitätsstufe = MQ100

446386-10G
SIAL446386-10GEA 284 EUR
SIAL446386-10G
2-(p-Toluolsulfonyl)ethanol, Sigma-Aldrich®
2-(p-Toluolsulfonyl)ethanol
Formel: C₉H₁₂O₃S
Molecular Weight: 200,2600 g/mol
MDL: MFCD00041757
CAS-Nummer: 22381-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR