Diethylethyl(phenyl)malonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diethyl 2-ethyl-2-phenylmalonate

M-Clarity™ Qualitätsstufe = MQ100

446416-5ML
SIAL446416-5MLEA 167 EUR
SIAL446416-5ML
Diethylethyl(phenyl)malonat, Sigma-Aldrich®
Diethylethyl(phenyl)malonat
Formel: C₁₅H₂₀O₄
Molecular Weight: 264,32 g/mol
MDL: MFCD00040759
CAS-Nummer: 76-67-5
EINECS: 200-978-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR