6-Fluorsalicylsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Fluor-2-hydroxybenzoesäure

M-Clarity™ Qualitätsstufe = MQ100

446505-1G
SIAL446505-1GEA 139 EUR
SIAL446505-1G
6-Fluorsalicylsäure, Sigma-Aldrich®
6-Fluorsalicylsäure
Formel: C₇H₅FO₃
Molecular Weight: 156,11 g/mol
MDL: MFCD00052179
CAS-Nummer: 67531-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR