(2,3,6-Trifluorphenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-(2,3,6-Trifluorophenyl)acetic acid

M-Clarity™ Qualitätsstufe = MQ200

446572-1G
SIAL446572-1GEA 73.1 EUR
SIAL446572-1G
(2,3,6-Trifluorphenyl)essigsäure, Sigma-Aldrich®
(2,3,6-Trifluorphenyl)essigsäure
Formel: C₆H₂F₃CH₂COOH
Molecular Weight: 190,12 g/mol
MDL: MFCD00061217
CAS-Nummer: 114152-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR