1H,1H-Heptadecafluor-1-nonanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1H,1H-Perfluor-1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Heptadecafluor-1-nonanol

M-Clarity™ Qualitätsstufe = MQ100

446823-5G
SIAL446823-5GEA 47.2 EUR
SIAL446823-5G
1H,1H-Heptadecafluor-1-nonanol, Sigma-Aldrich®
1H,1H-Heptadecafluor-1-nonanol
Formel: C₉H₃F₁₇O
Molecular Weight: 450,1 g/mol
MDL: MFCD00153183
CAS-Nummer: 423-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR