1H,1H-Heneicosafluor-1-undecanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1H,1H-Perfluor-1-undecanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heneicosafluor-1-undecanol

M-Clarity™ Qualitätsstufe = MQ100

446858-5G
SIAL446858-5GEA 67.6 EUR
SIAL446858-5G
1H,1H-Heneicosafluor-1-undecanol, Sigma-Aldrich®
1H,1H-Heneicosafluor-1-undecanol
Formel: C₁₁H₃F₂₁O
Molecular Weight: 550,11 g/mol
MDL: MFCD00153269
CAS-Nummer: 307-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR