N,N'-Bis(tert-butyl)ethylendiamin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

447005-25ML
SIAL447005-25MLEA 128 EUR
SIAL447005-25ML
N,N'-Bis(tert-butyl)ethylendiamin, Sigma-Aldrich®
N,N'-Bis(tert-butyl)ethylendiamin
Formel: C₁₀H₂₄N₂
MDL: MFCD00014996
CAS-Nummer: 4062-60-6
EINECS: 223-769-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR