Borwasserstoff-Dimethylsulfid-Komplex 5 mol/l in Diethylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Borane-methyl sulphide complex

M-Clarity™ Qualitätsstufe = MQ100

447145-100ML
SIAL447145-100MLEA 97 EUR
SIAL447145-100ML
Borwasserstoff-Dimethylsulfid-Komplex 5 mol/l in Diethylether, Sigma-Aldrich®
Borwasserstoff-Dimethylsulfid-Komplex
Formel: C₂H₉BS
Molecular Weight: 75,97 g/mol
MDL: MFCD00013189
CAS-Nummer: 13292-87-0
EINECS: 236-313-6
UN: 2924
ADR: 3,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR