Germanium(IV)isopropoxid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Germaniumtetraisopropoxid, Germanium(IV)isopropylat, Tetraisopropylorthogermanat, Germaniumtetraisopropanolat, Germaniumtetraisopropylat, Germanium(IV)isopropanolat

M-Clarity™ Qualitätsstufe = MQ200

447641-5G
SIAL447641-5GEA 252 EUR
SIAL447641-5G
Germanium(IV)isopropoxid, Sigma-Aldrich®
Germanium(IV)isopropoxid
Formel: C₁₂H₂₈GeO₄
Molecular Weight: 308,96 g/mol
MDL: MFCD00015339
CAS-Nummer: 21154-48-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR