(R)-(-)-3-Isopropyl-2,5-piperazindion, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-3-Isopropylpiperazine-2,5-dione

M-Clarity™ Qualitätsstufe = MQ100

448362-1G
SIAL448362-1GEA 26.4 EUR
SIAL448362-1G
(R)-(-)-3-Isopropyl-2,5-piperazindion, Sigma-Aldrich®
(R)-(-)-3-Isopropyl-2,5-piperazindion
Formel: C₇H₁₂N₂O₂
Molecular Weight: 156,18 g/mol
MDL: MFCD00210060
CAS-Nummer: 143673-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR