Diethoxymethylphenylsilan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diethoxy(methyl)(phenyl)silane

M-Clarity™ Qualitätsstufe = MQ100

448605-10ML
SIAL448605-10MLEA 72.6 EUR
SIAL448605-10ML
Diethoxymethylphenylsilan, Sigma-Aldrich®
Diethoxymethylphenylsilan
Formel: C₁₁H₁₈O₂Si
Molecular Weight: 210,35 g/mol
MDL: MFCD00048541
CAS-Nummer: 775-56-4
EINECS: 212-275-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR